Learn More
Lenz Laborglasintrumente 2-Neck Adapters
Click to view available options
Standard Taper Joint No.:
Socket Vertical NS 14/23
Socket Vertical NS 19/26
Socket Vertical NS 29/32
Unit Size:
Each
Description
- With two NS sockets and one NS cone
- Ascending angle: 105°
- Made of DURAN™ tubing
Specifications
Specifications
| Material | Duran Glass |
| Standard Taper Joint No. | Socket Vertical NS 19/26 |