Learn More
Lenz Laborglasintrumente™ 2-Neck Adapters
Click to view available options
Cone Size:
NS 24/29
NS 29/32
Standard Taper Joint No.:
NS 14/23
NS 24/29
Unit Size:
Each
Description
- Acc. to DIN 12594
- Made of DURAN™ tubing
Specifications
Specifications
| Cone Size | NS 29/32 |
| Material | Duran Tubing |
| Quantity | 1 |
| Standard Taper Joint No. | NS 14/23 |