Learn More
(2-Naphthyloxy)acetyl Chloride, TRC
Product Code. 30475344
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30475344 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30475344 Supplier Toronto Research Chemicals Supplier No. TRCN3786551G
 Restricted Item
Estimated Shipment: 07-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2-Naphthyloxy)acetyl Chloride
CAS 40926-77-0
Molecular Formula C12 H9 Cl O2
Quantity 1 g
Synonym 2-(2-Naphthalenyloxy)acetyl Chloride, 2-(2-Naphthyloxy)acetyl Chloride, β-Naphthyloxyacetyl Chloride
InChI Formula InChI=1S/C12H9ClO2/c13-12(14)8-15-11-6-5-9-3-1-2-4-10(9)7-11/h1-7H,8H2
SMILES ClC(=O)COc1ccc2ccccc2c1
IUPAC Name 2-naphthalen-2-yloxyacetyl chloride
Molecular Weight (g/mol) 220.652
Formula Weight 220.029