Learn More
2-Naphthaldehyde, 98%
Product Code. 11466237
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11466237 5 g 5g
11476237 25 g 25g
11486237 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11466237 Supplier Thermo Scientific Alfa Aesar Supplier No. B25157.06
Request Bulk or Custom Format
CAS: 66-99-9 | C11H8O | 156.184 g/mol
2-Naphthaldehyde aids in stereo-selective formation of bicyclo 2-siloxy-2-alkoxyoxetanes. It is also used as catalytic agent and petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Naphthaldehyde aids in stereo-selective formation of bicyclo 2-siloxy-2-alkoxyoxetanes. It is also used as catalytic agent and petrochemical additive.
Solubility
Soluble in hot water.
Notes
Store at -20°C. Air Sensitive. Store away from string bases and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66-99-9
Molecular Formula C11H8O
Molecular Weight (g/mol) 156.184
MDL Number MFCD00004094
InChI Key PJKVFARRVXDXAD-UHFFFAOYSA-N
Synonym 2-naphthaldehyde, 2-naphthalenecarboxaldehyde, 2-formylnaphthalene, beta-naphthaldehyde, beta-formylnaphthalene, beta-naphthylaldehyde, beta-naphthylcarboxaldehyde, .beta.-naphthaldehyde, .beta.-naphthylaldehyde, .beta.-formylnaphthalene
PubChem CID 6201
ChEBI CHEBI:52368
IUPAC Name naphthalene-2-carbaldehyde
SMILES C1=CC=C2C=C(C=CC2=C1)C=O
Specifications
Melting Point 59°C to 62°C
Boiling Point 160°C (19 mmHg)
Quantity 5 g
Beilstein 507750
Sensitivity Air Sensitive
Solubility Information Soluble in hot water.
Formula Weight 156.18
Percent Purity 98%
Chemical Name or Material 2-Naphthaldehyde