Learn More
2-(Methylthio)pyridine-3-boronic acid, 95%, Thermo Scientific™
Product Code. 15423627
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15423627
Brand: Thermo Scientific Alfa Aesar H27162.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1072944-21-8
Molecular Formula C6H8BNO2S
Molecular Weight (g/mol) 169.005
MDL Number MFCD09037488
InChI Key QLZLLRKAZYHWTE-UHFFFAOYSA-N
Synonym 2-methylthio pyridine-3-boronic acid, 2-methylthio pyridin-3-yl boronic acid, 2-methylsulfanyl pyridin-3-yl boronic acid, 2-methylsulfanyl pyridine-3-boronic acid, 2-methylsulfanyl pyridin-3-ylboronic acid, 2-methylthio-3-pyridinyl boronic acid, acmc-2098of, boronic acid, b-2-methylthio-3-pyridinyl
PubChem CID 44755183
IUPAC Name (2-methylsulfanylpyridin-3-yl)boronic acid
SMILES B(C1=C(N=CC=C1)SC)(O)O
Specifications
Melting Point ∽100°C (decomposition)
Quantity 250 mg
Formula Weight 169.01
Percent Purity 95%
Chemical Name or Material 2-(Methylthio)pyridine-3-boronic acid