missing translation for 'onlineSavingsMsg'
Learn More

2-Methyltetrahydrofuran, Sigma-Aldrich™

Product Code. 15861381
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15861381 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15861381 Supplier Sigma Aldrich Supplier No. 8210930250
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Stabilized for synthesis

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -136°C
CAS 96-47-9
Density 0.86 g/mL at 25°C
Quantity 250 mL
MDL Number MFCD00005367
Synonym 2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan, 2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan
Boiling Point 78°C to 80°C
Molecular Weight (g/mol) 86.13
Grade Research
Chemical Name or Material 2-Methyltetrahydrofuran
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.