Learn More
2-Methylhexanoic acid, TRC
Specifications
Specifications
| CAS | 4536-23-6 |
| Molecular Formula | C7H14O2 |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C7H14O2/c1-3-4-5-6(2)7(8)9/h6H,3-5H2,1-2H3,(H,8,9) |
| SMILES | CCCCC(C)C(=O)O |
| IUPAC Name | 2-methylhexanoic acid |
| Molecular Weight (g/mol) | 130.19 |
| Formula Weight | 130.0994 |
| Chemical Name or Material | 2-Methylhexanoic acid |