Learn More
(+/-)-2-(Methylamino)succinic acid, monohydrate, TRC
Specifications
Specifications
| Chemical Name or Material | (+/-)-2-(Methylamino)succinic acid, monohydrate |
| CAS | 303750-06-3 |
| Molecular Formula | C5 H9 N O4 . H2 O |
| Quantity | 100 mg |
| InChI Formula | InChI=1S/C5H9NO4.H2O/c1-6-3(5(9)10)2-4(7)8;/h3,6H,2H2,1H3,(H,7,8)(H,9,10);1H2 |
| SMILES | O.CNC(CC(=O)O)C(=O)O |
| IUPAC Name | 2-(methylamino)butanedioic acid;hydrate |
| Molecular Weight (g/mol) | 165.144 |
| Formula Weight | 165.064 |