Learn More
2-(Methylamino)pyridine, 98%
Product Code. 11464140
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464140 25 g 25g
11445617 1 g 1g
11455617 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11464140 Supplier Thermo Scientific Alfa Aesar Supplier No. B23589.14
Request Bulk or Custom Format
CAS: 4597-87-9 | C6H8N2 | 108.14 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 4597-87-9
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00006250
InChI Key SVEUVITYHIHZQE-UHFFFAOYSA-N
Synonym 2-methylamino pyridine, 2-methylaminopyridine, 2-pyridinamine, n-methyl, methylamino-2 pyridine, n-2-pyridylmethylamine, pyridine, 2-methylamino, n-methyl-2-pyridinamine, 2-pyridinamine, n-methyl-9ci, n-methyl pyridimine
PubChem CID 20727
IUPAC Name N-methylpyridin-2-amine
SMILES CNC1=CC=CC=N1
Specifications
Melting Point 14°C to 15°C
Density 1.06
Boiling Point 200°C to 201°C
Flash Point 87°C (188°F)
Refractive Index 1.578
Quantity 25 g
UN Number UN2810
Beilstein 108188
Formula Weight 108.14
Percent Purity 98%
Chemical Name or Material 2-(Methylamino)pyridine