Learn More
2-(Methylamino-d3)-3',4'-(methylenedioxy)butyrophenone Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | Butylone-d3 (hydrochloride) |
| CAS | 1231710-63-6 |
| Molecular Formula | C12 2H3 H12 N O3 . Cl H |
| Quantity | 1 mg |
| Synonym | 1-Butanone','1-(1,3-benzodioxol-5-yl)-2-(methyl-d3-amino)-','hydrochloride (1:1), 1-(1,3-Benzodioxol-5-yl)-2-(methyl-d3-amino)-1-butanone hydrochloride (1:1), Butylone-(N-methyl-D3) Hydrochloride, 2-(Methylamino-d3)-3',4'-(methylenedioxy)butyrophenone Hydrochloride |
| InChI Formula | InChI=1S/C12H15NO3.ClH/c1-3-9(13-2)12(14)8-4-5-10-11(6-8)16-7-15-10;/h4-6,9,13H,3,7H2,1-2H3;1H/i2D3; |
| SMILES | Cl.[2H]C([2H])([2H])NC(CC)C(=O)c1ccc2OCOc2c1 |
| IUPAC Name | 1-(1,3-benzodioxol-5-yl)-2-(trideuteriomethylamino)butan-1-one;hydrochloride |
| Molecular Weight (g/mol) | 260.73 |
| Formula Weight | 260.1007 |