Learn More
2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide, Mikromol™
Product Code. 30052506
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052506 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30052506 Supplier Mikromol Supplier No. MM0161.080025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide
CAS 151262-93-0
Analyte or Component Names 2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide
Shipping Condition Room Temperature
Molecular Formula C11 H11 F3 N2 O3
Certifications/Compliance ISO 9001
Synonym Flutamide Imp. F (EP), o-Flutamide, 2-Methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide, Flutamide Impurity F
InChI Formula InChI=1S/C11H11F3N2O3/c1-6(2)10(17)15-8-5-7(11(12,13)14)3-4-9(8)16(18)19/h3-6H,1-2H3,(H,15,17)
SMILES CC(C)C(=O)Nc1cc(ccc1[N+](=O)[O-])C(F)(F)F
IUPAC Name 2-methyl-N-[2-nitro-5-(trifluoromethyl)phenyl]propanamide
Molecular Weight (g/mol) 276.21
Quantity 25 mg
Formula Weight 276.0722 g/mol