Learn More
2-Methyl-d3-pyridine, TRC
Product Code. 30409746
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30409746 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30409746 Supplier Toronto Research Chemicals Supplier No. TRCP4371965MG
 Restricted Item
Estimated Shipment: 11-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-d3-pyridine
CAS 10259-19-5
Molecular Formula C6 D3 H4 N
Quantity 5 mg
Synonym Pyridine','2-(methyl-d3)- (9CI), 2-Picoline-α,α,α-d3 (7CI','8CI), 2-(Methyl-d3)pyridine, 2-Methylpyridine-d3, 2-Methylpyridine-D3 (methyl-D3), 2-Picoline-d3, 2-Methylpyridine-α,α,α-D3
InChI Formula InChI=1S/C6H7N/c1-6-4-2-3-5-7-6/h2-5H,1H3/i1D3
SMILES [2H]C([2H])([2H])c1ccccn1
IUPAC Name 2-(trideuteriomethyl)pyridine
Molecular Weight (g/mol) 96.145
Formula Weight 96.0767