Learn More
2-Methyl-5-isopropylaniline, Thermo Scientific Chemicals
Product Code. 16657012 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 16657012
Brand: Thermo Scientific Acros 248790050
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2051-53-8
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.24
MDL Number MFCD00044503
InChI Key YKWALWNGEXPARQ-UHFFFAOYSA-N
IUPAC Name 2-methyl-5-(propan-2-yl)aniline
SMILES CC(C)C1=CC=C(C)C(N)=C1
Specifications
Alpha Vector METHYLISOPROPYLANILINE
Color Brown to Colorless
Density 0.95
Infrared Spectrum Conforms
Packaging glass bottle
Refractive Index 1.5350 to 1.5450 (20°C, 589 nm)
Quantity 5 g
Sensitivity Air sensitive
Formula Weight 149.23
Physical Form Liquid
Chemical Name or Material 2-Methyl-5-isopropylaniline