missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-4-(trifluoromethoxy)phenylacetonitrile, 97%, Thermo Scientific™

Catalog Number 15468365
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
886763-09-3
C10H8F3NO
215.18
MFCD07368719
YQTMGMGJQACTFN-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy phenylacetonitrile, 2-2-methyl-4-trifluoromethoxy phenyl acetonitrile, 2-2-methyl-4-trifluoromethoxy phenyl ethanenitrile
2783306
2-[2-methyl-4-(trifluoromethoxy)phenyl]acetonitrile
CC1=C(CC#N)C=CC(OC(F)(F)F)=C1
This item is not returnable. View return policy
886763-09-3
C10H8F3NO
215.18
MFCD07368719
YQTMGMGJQACTFN-UHFFFAOYSA-N
2-methyl-4-trifluoromethoxy phenylacetonitrile, 2-2-methyl-4-trifluoromethoxy phenyl acetonitrile, 2-2-methyl-4-trifluoromethoxy phenyl ethanenitrile
2783306
2-[2-methyl-4-(trifluoromethoxy)phenyl]acetonitrile
CC1=C(CC#N)C=CC(OC(F)(F)F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3439
Formula Weight 215.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-4-(trifluoromethoxy)phenylacetonitrile
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'