Learn More
2-Methyl-3-(2’,3’-dihydroxypropyl)quinoxaline, TRC
Product Code. 30484269
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30484269 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30484269 Supplier Toronto Research Chemicals Supplier No. TRCM302225250MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Methyl-3-(2',3'-dihydroxypropyl)quinoxaline
CAS 42015-36-1
Molecular Formula C12 H14 N2 O2
Quantity 250 mg
Synonym 1,2-Propanediol','3-(3-methyl-2-quinoxalinyl)-, 3-(3-Methyl-2-quinoxalinyl)-1,2-propanediol, 2-Methyl-3-(2',3'-dihydroxypropyl)quinoxaline
InChI Formula InChI=1S/C12H14N2O2/c1-8-12(6-9(16)7-15)14-11-5-3-2-4-10(11)13-8/h2-5,9,15-16H,6-7H2,1H3
SMILES Cc1nc2ccccc2nc1CC(O)CO
IUPAC Name 3-(3-methylquinoxalin-2-yl)propane-1,2-diol
Molecular Weight (g/mol) 218.25
Formula Weight 218.1055