Learn More
2-Methyl-1H-imidazole-4-carbothioamide, 97%, Thermo Scientific™
Product Code. 10009471
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10009471 250 mg 250mg
10652073 1 g 1g
11360747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10009471 Supplier Thermo Scientific Maybridge Supplier No. SB02117CB
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Chemical Identifiers
CAS 129486-91-5
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.192
MDL Number MFCD03659724
InChI Key MNYPQSNAWPZXRV-UHFFFAOYSA-N
Synonym 2-methyl-1h-imidazole-4-carbothioamide, 1h-imidazole-5-carbothioamide,2-methyl, 1h-imidazole-5-carbothioamide, 2-methyl, acmc-1c27g, amino 2-methylimidazol-4-yl methane-1-thione, 2-methyl-3h-imidazole-4-carbothioamide
PubChem CID 2796714
IUPAC Name 2-methyl-1H-imidazole-5-carbothioamide
SMILES CC1=NC=C(N1)C(=S)N
Specifications
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 141.2
Percent Purity 97%
Chemical Name or Material 2-Methyl-1H-imidazole-4-carbothioamide