Learn More
2-Methyl-1-[(3-nitro-4-quinolinyl)amino]-2-propanol, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methyl-1-[(3-nitro-4-quinolinyl)amino]-2-propanol |
| CAS | 129655-57-8 |
| Molecular Formula | C13H15N3O3 |
| Quantity | 250 mg |
| Synonym | 2-Methyl-1-[(3-nitroquinolin-4-yl)amino]propan-2-ol, N-(2-Hydroxy-2-methylpropyl)-3-nitroquinolin-4-amine |
| InChI Formula | InChI=1S/C13H15N3O3/c1-13(2,17)8-15-12-9-5-3-4-6-10(9)14-7-11(12)16(18)19/h3-7,17H,8H2,1-2H3,(H,14,15) |
| SMILES | [O-][N+](C1=CN=C2C=CC=CC2=C1NCC(C)(C)O)=O |
| IUPAC Name | 2-methyl-1-((3-nitroquinolin-4-yl)amino)propan-2-ol |
| Molecular Weight (g/mol) | 261.28 |
| Formula Weight | 261.11 |