Learn More
2-(Methoxymethoxy)-1,3-dimethylbenzene-13C8,D3, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Methoxymethoxy)-1,3-dimethylbenzene-13C8,D3 |
| Molecular Formula | 13C8 C2 D3 H11 O2 |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C10H14O2/c1-8-5-4-6-9(2)10(8)12-7-11-3/h4-6H,7H2,1-3H3/i1+1,2+1D3,4+1,5+1,6+1,8+1,9+1,10+1 |
| SMILES | [2H][13C]([2H])([2H])[13c]1[13cH][13cH][13cH][13c]([13CH3])[13c]1OCOC |
| IUPAC Name | 2-(methoxymethoxy)-1-(113C)methyl-3-(trideuterio(113C)methyl)(1,2,3,4,5,6-13C6)cyclohexa-1,3,5-triene |
| Molecular Weight (g/mol) | 177.177 |
| Formula Weight | 177.145 |