Learn More
2-Methoxyisonicotinic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 105596-63-2 |
| Molecular Formula | C7H7NO3 |
| MDL Number | MFCD04115718 |
| Quantity | 500 g |
| SMILES | O=C(O)C1=CC=NC(OC)=C1 |
| Molecular Weight (g/mol) | 153.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Methoxyisonicotinic acid |