missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15460226
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15460226 250 mg 250mg
15450226 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15460226

Brand: Thermo Scientific Alfa Aesar H52932.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 240139-82-6
Molecular Formula C8H8BF3O3
Molecular Weight (g/mol) 219.954
MDL Number MFCD04039222
InChI Key PXDQUMRSSQYJSX-UHFFFAOYSA-N
Synonym 2-methoxy-5-trifluoromethylphenylboronic acid, 2-methoxy-5-trifluoromethyl phenylboronic acid, 2-methoxy-5-trifluoromethyl benzeneboronic acid, 2-methoxy-5-trifluoromethyl phenyl boronic acid, 5-trifluoromethyl-2-methoxyphenylboronic acid, 2-methoxy-5-trifluoromethylphenyl boronic acid, boronic acid,b-2-methoxy-5-trifluoromethyl phenyl, b-2-methoxy-5-trifluoromethyl phenyl-boronic acid, boronic acid, b-2-methoxy-5-trifluoromethyl phenyl
PubChem CID 2782676
IUPAC Name [2-methoxy-5-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(F)(F)F)OC)(O)O

Specifications

Melting Point 144°C to 148°C
Quantity 250 mg
Formula Weight 219.95
Percent Purity 98%
Chemical Name or Material 2-Methoxy-5-(trifluoromethyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.