Learn More
2-Methoxy-5-nitroformanilide, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Methoxy-5-nitroformanilide |
| CAS | 149686-06-6 |
| Molecular Formula | C8 H8 N2 O4 |
| Quantity | 5 g |
| InChI Formula | InChI=1S/C8H8N2O4/c1-14-8-3-2-6(10(12)13)4-7(8)9-5-11/h2-5H,1H3,(H,9,11) |
| SMILES | COc1ccc(cc1NC=O)[N+](=O)[O-] |
| IUPAC Name | N-(2-methoxy-5-nitrophenyl)formamide |
| Molecular Weight (g/mol) | 196.16 |
| Formula Weight | 196.0484 |