missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-4-(trifluoromethoxy)benzyl bromide, 97%, Thermo Scientific™

Catalog Number 11396849
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
886500-35-2
C9H8BrF3O2
285.06
MFCD04115970
JNWHGFGOMOVIMV-UHFFFAOYSA-N
2-methoxy-4-trifluoromethoxy benzyl bromide, 1-bromomethyl-2-methoxy-4-trifluoromethoxy benzene
5005442
1-(bromomethyl)-2-methoxy-4-(trifluoromethoxy)benzene
COC1=C(CBr)C=CC(OC(F)(F)F)=C1
This item is not returnable. View return policy
886500-35-2
C9H8BrF3O2
285.06
MFCD04115970
JNWHGFGOMOVIMV-UHFFFAOYSA-N
2-methoxy-4-trifluoromethoxy benzyl bromide, 1-bromomethyl-2-methoxy-4-trifluoromethoxy benzene
5005442
1-(bromomethyl)-2-methoxy-4-(trifluoromethoxy)benzene
COC1=C(CBr)C=CC(OC(F)(F)F)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Formula Weight 285.06
Percent Purity 97%
Chemical Name or Material 2-Methoxy-4-(trifluoromethoxy)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'