Learn More
2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester, 96%, Thermo Scientific™
Product Code. 15401786
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15401786
Brand: Thermo Scientific Alfa Aesar H54933.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1083168-94-8
Molecular Formula C12H17BN2O5
Molecular Weight (g/mol) 280.087
MDL Number MFCD12923397
InChI Key XWXOPFXUTOQOSM-UHFFFAOYSA-N
Synonym 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-pyridine, 2-methoxy-3-nitropyridine-5-boronic acid pinacol ester, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-3-nitropyridine, 6-methoxy-5-nitropyridine-3-boronic acid pinacol ester, 2-methoxy-3-nitro-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-methoxy-3-nitro-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, acmc-2098zq, 2-methoxy-3-nitropyridin-5-ylboronic acid pinacol ester, 6-methoxy-5-nitropyridin-3-yl boronic acid pinacol ester
PubChem CID 57416495
IUPAC Name 2-methoxy-3-nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=C(N=C2)OC)[N+](=O)[O-]
Specifications
Melting Point 118°C to 119°C
Quantity 250 mg
Formula Weight 280.08
Percent Purity 96%
Chemical Name or Material 2-Methoxy-3-nitropyridine-5-boronic acid pinacol ester