Learn More
2-Mercaptoethanol, ≥98%, MP Biomedicals™
Product Code. 11471711 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
250 mL
500 mL
1 L
Unit Size:
1 L
100mL
250mL
500mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471711 1 L 1 L
11441711 100 mL 100mL
11451711 250 mL 250mL
11461711 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11471711 Supplier MP Biomedicals Supplier No. 0219024291
  • ONLINE EXCLUSIVE PRICE
    1900.00 NOK
    1808.23 NOK / 1 L
    Save 91.77 NOK
    4% Off
 Restricted Item
Estimated Shipment: 19-01-2027
Chemical Identifiers
CAS 60-24-2
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00004890
InChI Key DGVVWUTYPXICAM-UHFFFAOYSA-N
Synonym 2-mercaptoethanol, mercaptoethanol, thioglycol, beta-mercaptoethanol, ethanol, 2-mercapto, 2-thioethanol, 2-hydroxy-1-ethanethiol, thioethylene glycol, 2-hydroxyethanethiol, 2-hydroxyethyl mercaptan
PubChem CID 1567
ChEBI CHEBI:41218
SMILES OCCS
Specifications
Color Colorless to Yellow
Density 1.12g/cm3 at 20°C
Assay Percent Range ≥98%
Quantity 1 L
IUPAC Name 2-sulfanylethan-1-ol
Formula Weight 78.13
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethanol