Learn More
2-Mercaptobenzoic Acid-d4, TRC
Product Code. 30481441
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481441 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30481441 Supplier Toronto Research Chemicals Supplier No. TRCM251702250MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Mercaptobenzoic Acid-d4
CAS 1246817-70-8
Molecular Formula C7H2D4O2S
Quantity 250 mg
Synonym 2-Thiosalicylic Acid-d4, 2-Carboxybenzenethiol-d4, 2-Carboxythiophenol-d4, 2-Sulfanylbenzoic Acid-d4, Thiophenol-2-carboxylic Acid-d4, Thiosalicylic Acid-d4, o-Carboxythiophenol-d4, o-Sulfhydrylbenzoic Acid-d4, NSC 2184-d4, NSC 660640-d4
InChI Formula InChI=1S/C7H6O2S/c8-7(9)5-3-1-2-4-6(5)10/h1-4,10H,(H,8,9)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c(C(=O)O)c(S)c1[2H]
IUPAC Name 2,3,4,5-tetradeuterio-6-sulfanylbenzoic acid
Molecular Weight (g/mol) 158.21
Formula Weight 158.034