Learn More
2-Mercaptobenzoic Acid-d4, TRC
Product Code. 30483230
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30483230 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30483230 Supplier Toronto Research Chemicals Supplier No. TRCM25170225MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Mercaptobenzoic Acid-d4
CAS 1246817-70-8
Molecular Formula C7H2D4O2S
Quantity 25 mg
Synonym 2-Thiosalicylic Acid-d4, 2-Carboxybenzenethiol-d4, 2-Carboxythiophenol-d4, 2-Sulfanylbenzoic Acid-d4, Thiophenol-2-carboxylic Acid-d4, Thiosalicylic Acid-d4, o-Carboxythiophenol-d4, o-Sulfhydrylbenzoic Acid-d4, NSC 2184-d4, NSC 660640-d4
InChI Formula InChI=1S/C7H6O2S/c8-7(9)5-3-1-2-4-6(5)10/h1-4,10H,(H,8,9)/i1D,2D,3D,4D
SMILES [2H]c1c([2H])c([2H])c(C(=O)O)c(S)c1[2H]
IUPAC Name 2,3,4,5-tetradeuterio-6-sulfanylbenzoic acid
Molecular Weight (g/mol) 158.21
Formula Weight 158.034