missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Mercaptobenzimidazole, TRC
Specifications
Specifications
| Quantity | 1 g |
| Synonym | 1,3-Dihydro-2H-benzimidazole-2-thione, 2-Benzimidazolethiol (8CI), 2-Benzimidazolinethione (6CI,7CI), 1,3-Dihydro-2H-benzimidazole-2-thione, 1,3-Dihydro-2H-benzoimidazole-2-thione, 1H-Benzimidazole-2-thiol, 1H-Benzo[d]imidazole-2(3H)-thione, 1H-Benzo[d]imidazole-2-thiol, 2,3-Dihydro-1H-benzimidazole-2-thione, 2-Benzimidazolethione, 2-Mercapto-1H-benzimidazole, 2-Mercaptobenzimidazole, 2-Sulfhydrylbenzimidazole, 2-Thiobenzimidazole, AO-MB, ASM MB, Antage MB, Antigene MB, Antioxidant MB, Antioxidant MB/K, Benzimidazolin-2-thione, Curekind MBI/B, G 580, MB, MB/K, MBI 80, NSC 186246, NSC 21414, Nocrac MB, Nonflex MB, Permanax 21, Rhenogran MBI 80, Sandant MB, Sumilizer MB, Vulkanox MB, Vulkanox MBI, Vulkanox MG, o-Phenylenethiourea, 1H-Benzimidazole-2-thiol, Lansoprazole Imp. E (EP) |
| SMILES | Sc1nc2ccccc2[nH]1 |
| Formula Weight | 150.0252 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?