missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidin-4(3H)-one, 96%, Thermo Scientific™

Product Code. 15425185
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15425185 1 g 1g
15445185 250 mg 250mg
15435185 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15425185 Supplier Thermo Scientific Alfa Aesar Supplier No. H33976.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299919-85-0
Molecular Formula C17H16N2OS2
Molecular Weight (g/mol) 328.448
MDL Number MFCD01896130
InChI Key IVSIHDBMSUYSAH-UHFFFAOYSA-N
Synonym 2-mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydro-3h-benzo 4,5 thieno 2,3-d pyrimidin-4-one, 2-mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidin-4 3h-one, 7-methyl-3-phenyl-2-sulfanyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidin-4 3h-one, 2-mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidin-4 3h-one, 7-methyl-3-phenyl-2-thioxo-2,3,5,6,7,8-hexahydro 1 benzothieno 2,3-d pyrimidin-4 1h-one, 1 benzothieno 2,3-d pyrimidin-4 1h-one,2,3,5,6,7,8-hexahydro-7-methyl-3-phenyl-2-thioxo, 11-methyl-4-phenyl-5-sulfanylidene-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7-dien-3-one, 2-mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo b thieno 2,3-d pyrimidin-4 3h-one, 7-methyl-3-phenyl-2-thioxo-1,3,5,6,7,8-hexahydrobenzo b thiopheno 2,3-d pyrimi din-4-one, 1 benzothieno 2,3-d pyrimidin-4 3h-one, 5,6,7,8-tetrahydro-2-mercapto-7-methyl-3-phenyl
PubChem CID 2814607
IUPAC Name 7-methyl-3-phenyl-2-sulfanylidene-5,6,7,8-tetrahydro-1H-[1]benzothiolo[2,3-d]pyrimidin-4-one
SMILES CC1CCC2=C(C1)SC3=C2C(=O)N(C(=S)N3)C4=CC=CC=C4

Specifications

Melting Point 343°C to 345°C
Quantity 1 g
Formula Weight 328.46
Percent Purity 96%
Chemical Name or Material 2-Mercapto-7-methyl-3-phenyl-5,6,7,8-tetrahydrobenzo[b]thieno[2,3-d]pyrimidin-4(3H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.