Learn More
2-Mercapto-4,6-dimethylpyrimidine sodium salt, 98%, Thermo Scientific™
Product Code. 11411427
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411427 250 g 250g
11401427 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11411427 Supplier Thermo Scientific Alfa Aesar Supplier No. B24925.30
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41840-27-1
Molecular Formula C6H7N2NaS
Molecular Weight (g/mol) 162.186
MDL Number MFCD04039775
InChI Key HUDKHPCVAMVYEQ-UHFFFAOYSA-M
Synonym 4,6-dimethylpyrimidine-2-thiol sodium salt, 2-mercapto-4,6-dimethylpyrimidine sodium salt, 4,6-dimethyl-2-mercaptopyrimidine sodium salt, sodium 4,6-dimethylpyrimidin-2-yl sulfanide, 4,6-dimethyl-2-sodiothiopyrimidine, 4,6-dimethyl-2-sodiosulfanyl pyrimidine
PubChem CID 23669009
IUPAC Name sodium;4,6-dimethylpyrimidine-2-thiolate
SMILES CC1=CC(=NC(=N1)[S-])C.[Na+]
Specifications
Quantity 250 g
Formula Weight 162.18
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4,6-dimethylpyrimidine sodium salt