Specifications
| CAS | 109-56-8 |
| Molecular Formula | C5H13NO |
| Quantity | 10 g |
| Synonym | 2-isopropylamino ethanol, n-isopropylethanolamine, 2-isopropylaminoethanol, isopropylaminoethanol, ethanolisopropylamine, ethanol, 2-1-methylethyl amino, n-isopropylaminoethanol, monoisopropylaminoethanol, n-hydroxyethyl isopropylamine |
| InChI Key | RILLZYSZSDGYGV-UHFFFAOYSA-N |
| SMILES | CC(C)NCCO |
| IUPAC Name | 2-(propan-2-ylamino)ethanol |
| Molecular Weight (g/mol) | 103.165 |
| PubChem CID | 7994 |
| Formula Weight | 103.7 |
| Show More |