Learn More
Alfa Aesar™ 2-Isopropoxypyridine-3-boronic acid pinacol ester, 96%
Catalog Number 11414530
Click to view available options
Quantity:
1 g
5 g
848243-25-4
C14H22BNO3
263.144
MFCD07366722
DLFPLUKZBSELRD-UHFFFAOYSA-N
2-isopropoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-isopropoxypyridine-3-boronic acid pinacol ester, 2-isopropoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-isopropoxypyridin-3-boronic acid pinacol ester, 2-isopropoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-propan-2-yloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem16590, 2-isopropoxypyridin-3-yl boronic acid pinacol ester, 2-propan-2-yloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-1-methylethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
17750192
2-propan-2-yloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)OC(C)C
848243-25-4
C14H22BNO3
263.144
MFCD07366722
DLFPLUKZBSELRD-UHFFFAOYSA-N
2-isopropoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-isopropoxypyridine-3-boronic acid pinacol ester, 2-isopropoxy-3-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-isopropoxypyridin-3-boronic acid pinacol ester, 2-isopropoxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-propan-2-yloxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem16590, 2-isopropoxypyridin-3-yl boronic acid pinacol ester, 2-propan-2-yloxy-3-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-1-methylethoxy-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine
17750192
2-propan-2-yloxy-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
B1(OC(C(O1)(C)C)(C)C)C2=C(N=CC=C2)OC(C)C
Applications
2-Isopropoxypyridine-3-boronic acid pinacol ester is used as an intermediate in the pharmaceutical industry.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Quantity 1 g
Solubility Information Slightly miscible with water.
Formula Weight 263.15
Percent Purity 96%
Chemical Name or Material 2-Isopropoxypyridine-3-boronic acid pinacol ester