Learn More
2-Iodo-p-xylene, 98+%
Product Code. p-7043504
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
Product Code. Quantity unitSize
11324434 10 g 10g
11334434 50 g 50g
2 options
Product Code. 11324434
Brand: Thermo Scientific Alfa Aesar L01055.09
Request Bulk or Custom Format
CAS: 1122-42-5 | C8H9I | 232.064 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1122-42-5
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00013708
InChI Key WYZVNUSNUCABRF-UHFFFAOYSA-N
Synonym 2-iodo-p-xylene, 1,4-dimethyl-2-iodobenzene, p-xylene, 2-iodo, 1-iodo-2,5-dimethylbenzene, benzene, 2-iodo-1,4-dimethyl, 2-iodo-4-xylene, 2,5-dimethyliodobenzene, 2-iodo-1,4-dimethyl-benzene, 2,5-dimethyl-1-iodobenzene, pubchem3875
PubChem CID 70731
IUPAC Name 2-iodo-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)I
Specifications
Density 1.617
Boiling Point 229°C to 230°C
Quantity 10 g
Refractive Index 1.598
Beilstein 1905728
Formula Weight 232.07
Percent Purity ≥98%
Chemical Name or Material 2-Iodo-p-xylene