missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-(trifluoromethyl)pyridine, 96%, Thermo Scientific Chemicals

Catalog Number 15490769
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
100366-75-4
C6H3F3IN
273.00
MFCD08741351
WSEVWIIOEQZEOU-UHFFFAOYSA-N
22147841
2-iodo-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CN=C(I)C=C1
This item is not returnable. View return policy
100366-75-4
C6H3F3IN
273.00
MFCD08741351
WSEVWIIOEQZEOU-UHFFFAOYSA-N
22147841
2-iodo-5-(trifluoromethyl)pyridine
FC(F)(F)C1=CN=C(I)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Sensitivity Light sensitive
Synonym 2-iodo-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-iodopyridine, 2-iodo-5-trifluoromethyl-pyridine, pyridine, 2-iodo-5-trifluoromethyl, pubchem16308, acmc-1c5wh, 2-iodanyl-5-trifluoromethyl pyridine, pyridine,2-iodo-5-trifluoromethyl
Formula Weight 272.99
Percent Purity 96%
Chemical Name or Material 2-Iodo-5-(trifluoromethyl)pyridine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'