Learn More
2-Imidazolidinon 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30324245
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
Product Code. Quantity unitSize
30324245 5 mL 5mL
1 options
Product Code. 30324245
Brand: Dr. Ehrenstorfer DREV14284000AL100
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Imidazolidinon
CAS 120-93-4
Analyte or Component Names 2-Imidazolidinon
Shipping Condition +20°C
Molecular Formula C3 H6 N2 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
InChI Formula InChI=1S/C3H6N2O/c6-3-4-1-2-5-3/h1-2H2,(H2,4,5,6)
SMILES O=C1NCCN1
IUPAC Name imidazolidin-2-one
Solvent or Matrix Acetonitrile
Industry Type Food Testing
Molecular Weight (g/mol) 86.09
Quantity 5 mL
Formula Weight 86.048 g/mol