missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyquinoxaline, 98%, Thermo Scientific™

Catalog Number 15279936
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
1196-57-2
C8H6N2O
146.15
MFCD00006722
FFRYUAVNPBUEIC-UHFFFAOYSA-N
2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
14526
CHEBI:38890
1H-quinoxalin-2-one
O=C1NC2=CC=CC=C2N=C1
This item is not returnable. View return policy
1196-57-2
C8H6N2O
146.15
MFCD00006722
FFRYUAVNPBUEIC-UHFFFAOYSA-N
2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
14526
CHEBI:38890
1H-quinoxalin-2-one
O=C1NC2=CC=CC=C2N=C1
Melting Point 271.0°C to 272.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 100 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hot methanol
Formula Weight 146.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxyquinoxaline
compliance-icons
Product Identifier
  • 2-Hydroxyquinoxaline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'