Learn More
2-(Hydroxymethyl)pyridine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-(Hydroxymethyl)pyridine |
| CAS | 586-98-1 |
| Molecular Formula | C6H7NO |
| Quantity | 50 g |
| Synonym | 2-Pyridinylcarbinol, 2-Pyridinylmethanol, 2-Pyridylcarbinol, 2-Pyridylmethanol, Piconol, Pyridine-2-carbinol, α-Picolyl alcohol, 2-Pyridinemethanol |
| InChI Formula | InChI=1S/C6H7NO/c8-5-6-3-1-2-4-7-6/h1-4,8H,5H2 |
| SMILES | OCc1ccccn1 |
| IUPAC Name | pyridin-2-ylmethanol |
| Molecular Weight (g/mol) | 109.13 |
| Formula Weight | 109.0528 |