missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethylhydrazine, TRC

Product Code. 30401742
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 30401742

Brand: Toronto Research Chemicals TRCH9421801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxyethylhydrazine
CAS 109-84-2
Molecular Formula C2 H8 N2 O
Quantity 1 g
Synonym (2-Hydroxyethyl)hydrazine, (β-Hydroxyethyl)hydrazine, 1-(2-Hydroxyethyl)hydrazine, 2-Hydrazineethanol, 2-Hydrazinoethanol, 2-Hydrazinoethyl Alcohol, 2-Hydrazinylethanol, BOH, Ethanolhydrazine, HEH, Hydrazineethanol, Hydroxyethylhydrazine, N-(2-Hydroxyethyl)hydrazine, NSC 189820, 2-Hydrazino-ethanol
InChI Formula InChI=1S/C2H8N2O/c3-4-1-2-5/h4-5H,1-3H2
SMILES NNCCO
IUPAC Name 2-hydrazinylethanol
Molecular Weight (g/mol) 76.1
Formula Weight 76.0637
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.