Learn More
2-Hydroxy Nicotinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy Nicotinic Acid |
| CAS | 609-71-2 |
| Molecular Formula | C6H5NO3 |
| Quantity | 1 g |
| Synonym | 1,2-Dihydro-2-oxo-3-pyridinecarboxylic Acid, 1,2-Dihydro-2-oxonicotinic Acid 1,2-Dihydro-2-oxo-3-pyridinecarboxylic Acid, 1,2-Dihydro-2-oxonicotinic Acid, 2-Hydroxy-3-carboxypyridine, 2-Hydroxynicotinic Acid, 2-Hydroxypyridine-3-carboxylic Acid, 2-Oxo-1,2-dihydropyridine-3-carboxylic Acid, 3-Carboxy-2-pyridone, NSC 226152 |
| InChI Formula | InChI=1S/C6H5NO3/c8-5-4(6(9)10)2-1-3-7-5/h1-3H,(H,7,8)(H,9,10) |
| SMILES | OC(=O)c1cccnc1O |
| IUPAC Name | 2-hydroxypyridine-3-carboxylic acid |
| Molecular Weight (g/mol) | 139.11 |
| Formula Weight | 139.0269 |