Learn More
2-Hydroxy-5-methanesulfonamidobenzoic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxy-5-methanesulfonamidobenzoic Acid |
| CAS | 926243-08-5 |
| Molecular Formula | C8H9NO5S |
| Quantity | 1 g |
| Synonym | 2-hydroxy-5-(methylsulfonamido)benzoic acid, 2-Hydroxy-5-[(methylsulfonyl)amino]benzoic acid |
| InChI Formula | InChI=1S/C8H9NO5S/c1-15(13,14)9-5-2-3-7(10)6(4-5)8(11)12/h2-4,9-10H,1H3,(H,11,12) |
| SMILES | O=C(O)C1=CC(=CC=C1O)NS(=O)(=O)C |
| IUPAC Name | 2-hydroxy-5-(methylsulfonamido)benzoic acid |
| Molecular Weight (g/mol) | 231.22 |
| Formula Weight | 231.02 |