Learn More
2-Hydroxy-3-cyanopyridine, TRC
Product Code. 30406306
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406306 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30406306 Supplier Toronto Research Chemicals Supplier No. TRCH825140500MG
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-3-cyanopyridine
CAS 20577-27-9
Molecular Formula C6 H4 N2 O
Quantity 500 mg
InChI Formula InChI=1S/C6H4N2O/c7-4-5-2-1-3-8-6(5)9/h1-3H,(H,8,9)
SMILES Oc1ncccc1C#N
IUPAC Name 2-hydroxypyridine-3-carbonitrile
Molecular Weight (g/mol) 120.11
Formula Weight 120.0324