Learn More
2-Hydroxy-3-cyanopyridine, TRC
Product Code. 30397245
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30397245 Supplier Toronto Research Chemicals Supplier No. TRCH8251401G
 Restricted Item
Estimated Shipment: 01-10-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-3-cyanopyridine
CAS 20577-27-9
Molecular Formula C6 H4 N2 O
Quantity 1 g
InChI Formula InChI=1S/C6H4N2O/c7-4-5-2-1-3-8-6(5)9/h1-3H,(H,8,9)
SMILES Oc1ncccc1C#N
IUPAC Name 2-hydroxypyridine-3-carbonitrile
Molecular Weight (g/mol) 120.11
Formula Weight 120.0324