Learn More
2-Hydroxy-1-naphthaldehyde, tech.
Product Code. p-146503 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10347090 100 g 100g
10421163 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10347090 Supplier Thermo Scientific Acros Supplier No. 121601000
Add to basket
CAS: 708-06-5 | C11H8O2 | 172.18 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 708-06-5
MDL Number MFCD00004005
InChI Key NTCCNERMXRIPTR-UHFFFAOYSA-N
Synonym 2-hydroxy-1-naphthaldehyde, 1-formyl-2-naphthol, 2-hydroxy-1-naphthylaldehyde, 2-hydroxy-1-napthaldehyde, 2-hydroxynaphthaldehyde, 2-naphthol 1-carboxaldehyde, 2-hydroxy-1-naphthalaldehyde, 1-naphthalenecarboxaldehyde, 2-hydroxy, 2-hydroxy-1-naphthalenecarboxaldehyde, 1-hydroxy-2-naphthalenecarboxaldehyde
PubChem CID 12819
IUPAC Name 2-hydroxynaphthalene-1-carbaldehyde
SMILES C1=CC=C2C(=C1)C=CC(=C2C=O)O
Specifications
Melting Point >77.0°C
Color Yellow
Boiling Point 192.0°C (27.0 mmHg)
Infrared Spectrum Authentic
Packaging Plastic bottle
Molecular Formula C11H8O2
Linear Formula HOC10H6CHO
Quantity 100 g
Beilstein 08,143
Solubility Information Solubility in water: 40mg/l
Molecular Weight (g/mol) 172.18
Formula Weight 172.18
Percent Purity ≥93% (on dry substance)
Grade Technical
Physical Form Moist Crystalline Powder
Chemical Name or Material 2-Hydroxy-1-naphthaldehyde, tech.
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxy-1-naphthaldehyde
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.
Precautionary Statement