Learn More
2-Hydroxy-1-methyl-6-phenylimidazo(4,5-b)pyridine-d3, TRC
Product Code. 30388820
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30388820 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30388820 Supplier Toronto Research Chemicals Supplier No. TRCH79177710MG
 Restricted Item
Estimated Shipment: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Hydroxy-1-(trideuteromethyl)-6-phenylimidazo(4,5-b)pyridin
CAS 1020719-48-5
Molecular Formula C13 2H3 H8 N3 O
Quantity 10 mg
Synonym 2H-Imidazo[4,5-b]pyridin-2-one, 1,3-dihydro-1-(methyl-d3)-6-phenyl-, 1,3-Dihydro-1-trideuteriomethyl-6-phenyl-2H-imidazo[4,5-b]pyridin-2-one, 2-Hydroxy-1-methyl-6-phenylimidazo[4,5-b]pyridine-D3 (N-methyl-D3), 1-(Methyl-d3)-6-phenyl-1H-imidazo[4,5-b]pyridin-2-ol
InChI Formula InChI=1S/C13H11N3O/c1-16-11-7-10(9-5-3-2-4-6-9)8-14-12(11)15-13(16)17/h2-8H,1H3,(H,14,15,17)/i1D3
SMILES [2H]C([2H])([2H])n1c(O)nc2ncc(cc12)c3ccccc3
IUPAC Name 6-phenyl-1-(trideuteriomethyl)imidazo[4,5-b]pyridin-2-ol
Molecular Weight (g/mol) 228.26
Formula Weight 228.109