Learn More
2-Hydroxy-1,2-diphenylethanone oxime, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 441-38-3 |
| Molecular Formula | C14H13NO2 |
| MDL Number | MFCD00004501 |
| Quantity | 25 g |
| SMILES | OC(C1=CC=CC=C1)/C(C2=CC=CC=C2)=N/O |
| Molecular Weight (g/mol) | 227.26 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Hydroxy-1,2-diphenylethanone oxime |