Learn More
2-Hexynyl-5'-N-ethylcarboxamidoadenosine, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hexynyl-5'-N-ethylcarboxamidoadenosine |
| CAS | 141018-30-6 |
| Molecular Formula | C18H24N6O4 |
| Quantity | 5 mg |
| Synonym | 2-Hexynyl-5'-N-ethylcarboxamidoadenosine, 2-Hexynyl-NECA, HENECA |
| InChI Formula | InChI=1S/C18H24N6O4/c1-3-5-6-7-8-10-22-15(19)11-16(23-10)24(9-21-11)18-13(26)12(25)14(28-18)17(27)20-4-2/h9,12-14,18,25-26H,3-6H2,1-2H3,(H,20,27)(H2,19,22,23)/t12-,13+,14-,18+/m0/s1 |
| SMILES | CCCCC#Cc1nc(N)c2ncn([C@@H]3O[C@@H]([C@@H](O)[C@H]3O)C(=O)NCC)c2n1 |
| IUPAC Name | (2S,3S,4R,5R)-5-(6-amino-2-hex-1-ynylpurin-9-yl)-N-ethyl-3,4-dihydroxyoxolane-2-carboxamide |
| Molecular Weight (g/mol) | 388.42 |
| Formula Weight | 388.1859 |