missing translation for 'onlineSavingsMsg'
Learn More

2-(Fmoc-amino)-4-chlorobenzoic acid, 95%, Thermo Scientific™

Product Code. 11369785
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369785 5 g 5g
11359785 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369785 Supplier Thermo Scientific Alfa Aesar Supplier No. L19809.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 332121-92-3
Molecular Formula C22H16ClNO4
Molecular Weight (g/mol) 393.823
MDL Number MFCD02682157
InChI Key BIPQSYSWHUYDEA-UHFFFAOYSA-N
Synonym 2-9h-fluoren-9-yl methoxy carbonyl amino-4-chlorobenzoic acid, 2-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino benzoic acid, n-fmoc-4-chloroanthranilic acid, 2-n-fmoc-amino-4-chlorobenzoic acid, 4-chloro-2-fluoren-9-ylmethoxy carbonylamino benzoic acid, n-9-fluorenylmethoxycarbonyl-4-chloroanthranilic acid, 4-chloro-2-9h-fluoren-9-ylmethoxycarbonylamino-benzoic acid, benzoic acid, 4-chloro-2-9h-fluoren-9-ylmethoxy carbonyl amino
PubChem CID 7022993
IUPAC Name 4-chloro-2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=C(C=CC(=C4)Cl)C(=O)O

Specifications

Melting Point 218°C to 220°C (decomposition)
Quantity 5 g
Formula Weight 393.83
Percent Purity 95%
Chemical Name or Material 2-(Fmoc-amino)-4-chlorobenzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.