Learn More
2-Fluoroethanol, 95%
Product Code. 12694937 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
Product Code. 12694937
Brand: Thermo Scientific Acros 170830250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 371-62-0 | C2H5FO | 64.06 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 371-62-0
Molecular Formula C2H5FO
Molecular Weight (g/mol) 64.06
InChI Key GGDYAKVUZMZKRV-UHFFFAOYSA-N
Synonym ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
PubChem CID 9737
IUPAC Name 2-fluoroethanol
SMILES C(CF)O
Specifications
Melting Point -26.5°C
Density 1.1080g/mL
Boiling Point 100.0°C to 104.0°C
Flash Point 34°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula FCH2CH2OH
Quantity 25 g
Beilstein 01,I,17
Specific Gravity 1.108
Solubility Information Solubility in water: miscible
Formula Weight 64.06
Percent Purity 95%
Chemical Name or Material 2-Fluoroethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluoroethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /