Learn More
2-Fluoro-6-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
Product Code. 11430518
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430518 5 g 5g
11420518 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11430518 Supplier Thermo Scientific Alfa Aesar Supplier No. B24654.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 32890-94-1
Molecular Formula C8H3F4O2
Molecular Weight (g/mol) 207.10
MDL Number MFCD00040981
InChI Key LNARMXLVVGHCRP-UHFFFAOYSA-M
Synonym 2-fluoro-6-trifluoromethyl benzoic acid, otf-boa-6f, alpha,alpha,alpha,6-tetrafluoro-o-toluic acid, benzoic acid, 2-fluoro-6-trifluoromethyl, 6-fluoro-2-trifluoromethylbenzoic acid, 2-fluoro-6-trifluoromethyl-benzoic acid, 6-fluoro-2-trifluoromethyl benzoic acid, pubchem4759, acmc-20a40d, rarechem al bo 0626
PubChem CID 688123
SMILES [O-]C(=O)C1=C(F)C=CC=C1C(F)(F)F
Specifications
Melting Point 83°C to 88°C
Quantity 5 g
Beilstein 2112882
IUPAC Name 2-fluoro-6-(trifluoromethyl)benzoic acid
Formula Weight 208.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzoic acid