Learn More
2-Fluoro-4-(pentafluorothio)benzoic acid, 97%, Thermo Scientific™
Product Code. 15477107
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15477107 5 g 5g
15467107 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15477107 Supplier Thermo Scientific Alfa Aesar Supplier No. H34180.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1240257-03-7
Molecular Formula C7H4F6O2S
Molecular Weight (g/mol) 266.157
MDL Number MFCD16652503
InChI Key MGIDEXWQQLSCGW-UHFFFAOYSA-N
Synonym 2-fluoro-4-pentafluoro-??-sulfanyl benzoic acid, 2-fluoro-4-pentafluorothio benzoic acid, 2-fluoro-4-pentafluorosulfur benzoic acid
PubChem CID 66523491
IUPAC Name 2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)benzoic acid
SMILES C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)C(=O)O
Specifications
Quantity 5 g
Formula Weight 266.16
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-(pentafluorothio)benzoic acid