missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl salicylate, Thermo Scientific™

Product Code. p-7232423 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
This item is not returnable. View return policy

Product Code. 17342501

Brand: Thermo Scientific Acros 468830250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 118-60-5
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
MDL Number 00053300
InChI Key FMRHJJZUHUTGKE-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 2-hydroxybenzoate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1O

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Density 1.02
Flash Point 162°C
Physical Form Liquid
Color Colorless to Light Yellow
Melting Point -20°C
Boiling Point 190°C (21 mmHG)
Solubility Insoluble in water
Quantity 25 mL
Formula Weight 250.34
Chemical Name or Material 2-Ethylhexyl salicylate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.